Assay Details
Target Gene Details
Entrez Gene ID: | 85479 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C5 beta |
Gene Aliases: |
CSP-beta |
Location: |
Chr.8:66014983-66100520 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC5B | NM_033105.4 | NP_149096.2 | ||
| XM_011517618.2 | XP_011515920.1 | |||
| XM_011517620.2 | XP_011515922.1 | |||
| AF368276.1 | AAK60571.1 | |||
| AL133660.1 | CAB63773.2 | |||
| BC016742.2 | AAH16742.1 | |||
| BG701336.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73241293] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map