Assay Details
Target Gene Details
Entrez Gene ID: | 137682 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 6 |
Gene Aliases: |
C8orf38 |
Location: |
Chr.8:94895271-95118496 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF6 | XM_011516835.2 | XP_011515137.1 | ||
XM_017013028.1 | XP_016868517.1 | |||
XM_017013030.1 | XP_016868519.1 | |||
XM_017013031.1 | XP_016868520.1 | |||
XM_017013032.1 | XP_016868521.1 | |||
XM_017013035.1 | XP_016868524.1 | |||
XM_017013036.1 | XP_016868525.1 | |||
XM_017013039.1 | XP_016868528.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
