Assay Details
Target Gene Details
Entrez Gene ID: | 158399 |
Gene Name: | zinc finger protein 483 |
Gene Aliases: |
ZKSCAN16, ZSCAN48 |
Location: |
Chr.9:111525159-111577844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF483 | NM_001007169.3 | NP_001007170.1 | ||
| NM_133464.3 | NP_597721.2 | |||
| XM_017014337.1 | XP_016869826.1 | |||
| XM_017014338.1 | XP_016869827.1 | |||
| XM_017014339.1 | XP_016869828.1 | |||
| AB075842.1 | ||||
| AK092811.1 | BAC03981.1 | |||
| AK291267.1 | ||||
| BC065738.1 | AAH65738.1 | |||
| BC152427.1 | ||||
| BC152433.1 |
More Information
Set Membership: |
|

Genomic Map