Assay Details
Target Gene Details
Entrez Gene ID: | 286343 |
Gene Name: | leucine rich adaptor protein 1 like |
Gene Aliases: |
C9orf150, HYST0841, LRAP35b, bA3L8.2 |
Location: |
Chr.9:12774990-12823060 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LURAP1L | NM_203403.1 | NP_981948.1 | ||
| XM_005251443.2 | 2 | 1303 | XP_005251500.1 | |
| AB073606.1 | BAD38643.1 | |||
| AK074752.1 | ||||
| AK095824.1 | BAC04633.1 | |||
| BC036923.1 | AAH36923.1 | |||
| BM739457.1 |
Target Gene Details
Entrez Gene ID: | 101929467 |
Gene Name: | LURAP1L antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:12699999-12814389 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LURAP1L-AS1 | NR_125775.1 | |||
| BQ416907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891612 | Chr.9:12213879 - 13226946 on Build GRCh38 | Gain |
|
| nsv1021844 | Chr.9:12356017 - 13444976 on Build GRCh38 | Gain |
|
| esv2422212 | Chr.9:12448789 - 13127739 on Build GRCh38 | Duplication |
|
| esv2759671 | Chr.9:12235048 - 12784093 on Build GRCh38 | Loss |
|
| nsv1030835 | Chr.9:12717406 - 12840929 on Build GRCh38 | Gain |
|

Genomic Map