Assay Details
Target Gene Details
Entrez Gene ID: | 23071 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
PDIA10, TXNDC4 |
Location: |
Chr.9:99979179-100099052 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP44 | NM_015051.2 | NP_055866.1 | ||
| AB011145.1 | ||||
| AJ344330.2 | CAC87611.1 | |||
| AK075024.1 | ||||
| AY359048.1 | AAQ89407.1 | |||
| BC005374.1 | AAH05374.1 | |||
| BX648519.1 |
More Information
Set Membership: |
|

Genomic Map