Assay Details
Target Gene Details
Entrez Gene ID: | 51148 |
Gene Name: | cerebral endothelial cell adhesion molecule |
Gene Aliases: |
CEECAM1, GLT25D3 |
Location: |
Chr.9:128405993-128437351 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERCAM | NM_001286760.1 | NP_001273689.1 | ||
| NM_016174.4 | NP_057258.3 | |||
| XM_005252035.1 | XP_005252092.1 | |||
| XM_011518762.1 | XP_011517064.1 | |||
| XM_017014794.1 | XP_016870283.1 | |||
| AB040935.1 | 3 | 1671 | BAA96026.1 | |
| AK297929.1 | ||||
| AK314294.1 | ||||
| BC098432.1 | ||||
| BC108698.1 | AAI08699.1 | |||
| BC119698.1 | ||||
| BC119699.1 | ||||
| BX358794.2 | ||||
| CX871555.1 |
More Information
Set Membership: |
|

Genomic Map