Assay Details
Target Gene Details
Entrez Gene ID: | 100507299 |
Gene Name: | SMC5 antisense RNA 1 (head to head) |
Gene Aliases: |
MAMDC2-AS2 |
Location: |
Chr.9:70216059-70258874 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC5-AS1 | NR_039990.1 | 1 | 414 | |
| AK124136.1 | 1 | 406 | ||
| CR936772.1 | 1 | 398 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6555 | Chr.9:70225054 - 70259811 on Build GRCh38 | Insertion |
|
| nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain |
|
| nsv831615 | Chr.9:70237789 - 70404776 on Build GRCh38 | Gain |
|

Genomic Map