Assay Details
Target Gene Details
Entrez Gene ID: | 64768 |
Gene Name: | inositol-pentakisphosphate 2-kinase |
Gene Aliases: |
C9orf12, INSP5K2, IP5K, IPK1, bA476B13.1 |
Location: |
Chr.9:92613184-92670265 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPPK | NM_022755.5 | NP_073592.1 | ||
| XM_017015041.1 | XP_016870530.1 | |||
| AF351202.1 | AAK69692.1 | |||
| AF520811.1 | AAM75353.1 | |||
| AK023225.1 | BAB14476.1 | |||
| AK024267.1 | BAB14866.1 | |||
| AK123694.1 | ||||
| BC026154.2 | AAH26154.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115718412] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map