Assay Details
Target Gene Details
Entrez Gene ID: | 8513 |
Gene Name: | lipase F, gastric type |
Gene Aliases: |
GL, HGL, HLAL |
Location: |
Chr.10:88664389-88678815 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPF | NM_001198828.1 | NP_001185757.1 | ||
| NM_001198829.1 | NP_001185758.1 | |||
| NM_001198830.1 | NP_001185759.1 | |||
| NM_004190.3 | NP_004181.1 | |||
| XM_011540311.1 | XP_011538613.1 | |||
| A01046.1 | CAA00125.1 | |||
| AK223468.1 | BAD97188.1 | |||
| AK301310.1 | ||||
| AK301320.1 | ||||
| AK308224.1 | ||||
| AK312940.1 | ||||
| AL833751.1 | CAH56244.1 | |||
| BC112272.1 | AAI12273.1 | |||
| BC113711.1 | ||||
| X05997.1 | CAA29413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain |
|
| esv3624132 | Chr.10:88489037 - 88688695 on Build GRCh38 | Gain |
|
| esv3624134 | Chr.10:88555882 - 88703649 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map