Assay Details
Target Gene Details
Entrez Gene ID: | 79009 |
Gene Name: | DEAD-box helicase 50 |
Gene Aliases: |
GU2, GUB, RH-II/GuB, mcdrh |
Location: |
Chr.10:68901278-68956312 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX50 | NM_024045.1 | NP_076950.1 | ||
| XM_005270148.2 | XP_005270205.1 | |||
| XM_011540144.2 | XP_011538446.1 | |||
| XM_017016626.1 | XP_016872115.1 | |||
| XM_017016627.1 | XP_016872116.1 | |||
| XM_017016628.1 | XP_016872117.1 | |||
| XM_017016629.1 | XP_016872118.1 | |||
| XM_017016630.1 | XP_016872119.1 | |||
| AB051075.1 | ||||
| AF334103.1 | AAK29402.1 | |||
| AK223297.1 | BAD97017.1 | |||
| AK293577.1 | ||||
| AK301431.1 | ||||
| BC000272.1 | AAH00272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831907 | Chr.10:68871974 - 69029649 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map