Assay Details
Target Gene Details
Entrez Gene ID: | 9644 |
Gene Name: | SH3 and PX domains 2A |
Gene Aliases: |
FISH, SH3MD1, TKS5 |
Location: |
Chr.10:103594027-103855464 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3PXD2A | NM_014631.2 | NP_055446.2 | ||
| XM_005270294.4 | XP_005270351.1 | |||
| XM_005270295.4 | XP_005270352.1 | |||
| XM_006718079.3 | XP_006718142.1 | |||
| XM_011540385.2 | XP_011538687.1 | |||
| XM_017016969.1 | XP_016872458.1 | |||
| AK056469.1 | ||||
| CN307901.1 |
Target Gene Details
Entrez Gene ID: | 100505839 |
Gene Name: | SH3PXD2A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:103746779-103755409 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3PXD2A-AS1 | NR_038940.1 | |||
| AK056784.1 | ||||
| DA422325.1 |
More Information
Set Membership: |
|

Genomic Map