Assay Details
Target Gene Details
Entrez Gene ID: | 54764 |
Gene Name: | zinc finger RANBP2-type containing 1 |
Gene Aliases: |
TRABID |
Location: |
Chr.10:124916896-124988059 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB1 | XM_005269926.4 | XP_005269983.1 | ||
| XM_017016357.1 | XP_016871846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552289 | Chr.10:124875780 - 124998761 on Build GRCh38 | Loss |
|
| esv3624822 | Chr.10:124932102 - 124937760 on Build GRCh38 | Gain |
|
| nsv825605 | Chr.10:124936013 - 124936470 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map