Assay Details
Target Gene Details
Entrez Gene ID: | 9200 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 1 |
Gene Aliases: |
CAP, PTPLA |
Location: |
Chr.10:17589959-17617374 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACD1 | NM_014241.3 | NP_055056.3 | ||
| XM_005252641.4 | XP_005252698.1 | |||
| AF114494.1 | AAF21976.1 | |||
| BC010353.1 | AAH10353.1 |
More Information
Set Membership: |
|

Genomic Map