Assay Details
Target Gene Details
Entrez Gene ID: | 57209 |
Gene Name: | zinc finger protein 248 |
Gene Aliases: |
bA162G10.3 |
Location: |
Chr.10:37758452-37858202 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF248 | AK125538.1 | 1 | 1475 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622969 | Chr.10:37730948 - 37772865 on Build GRCh38 | Loss |
|
| nsv1040704 | Chr.10:37651096 - 38162989 on Build GRCh38 | Gain |
|
| dgv112e214 | Chr.10:37441894 - 37765337 on Build GRCh38 | Loss |
|

Genomic Map