Assay Details
Target Gene Details
Entrez Gene ID: | 8187 |
Gene Name: | zinc finger protein 239 |
Gene Aliases: |
HOK-2, MOK2 |
Location: |
Chr.10:43554516-43574619 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF239 | XM_005271832.2 | 2 | 2700 | XP_005271889.1 |
| XM_006718001.2 | 2 | 2799 | XP_006718064.1 | |
| XM_006718003.3 | 2 | 2699 | XP_006718066.1 | |
| XM_011540232.2 | 3 | 5786 | XP_011538534.1 | |
| XM_011540234.2 | 3 | 2833 | XP_011538536.1 | |
| XM_011540235.2 | 3 | 2832 | XP_011538537.1 | |
| XM_011540236.2 | 3 | 2841 | XP_011538538.1 | |
| XM_011540237.2 | 3 | 2791 | XP_011538539.1 | |
| XM_011540238.2 | 2 | 2643 | XP_011538540.1 | |
| XM_017016740.1 | 4 | 2870 | XP_016872229.1 |
More Information
Set Membership: |
|

Genomic Map