Assay Details
Target Gene Details
Entrez Gene ID: | 196740 |
Gene Name: | V-set and transmembrane domain containing 4 |
Gene Aliases: |
C10orf72 |
Location: |
Chr.10:49014245-49129118 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM4 | NM_001031746.4 | NP_001026916.2 | ||
| XM_017015825.1 | XP_016871314.1 | |||
| XM_017015826.1 | XP_016871315.1 | |||
| XM_017015827.1 | XP_016871316.1 | |||
| XM_017015828.1 | XP_016871317.1 | |||
| AK297931.1 | ||||
| BC041414.1 | AAH41414.1 | |||
| DA176757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623162 | Chr.10:49011836 - 49038527 on Build GRCh38 | Loss |
|
| nsv831866 | Chr.10:48957135 - 49127231 on Build GRCh38 | Gain+Loss |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79553893] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map