Assay Details
Target Gene Details
| Entrez Gene ID: | 1791 | 
| Gene Name: | DNA nucleotidylexotransferase | 
| Gene Aliases: | TDT | 
| Location: | Chr.10:96304328-96338564 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DNTT | NM_001017520.1 | NP_001017520.1 | ||
| NM_004088.3 | NP_004079.3 | |||
| AB046378.1 | BAB72001.1 | |||
| AK223317.1 | BAD97037.1 | |||
| AK313615.1 | ||||
| BC012920.1 | AAH12920.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2761620 | Chr.10:96287288 - 96329992 on Build GRCh38 | Gain |  DNTT | 
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |  DNTT  BLNK  TLL2  OPALIN | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top