Assay Details
Target Gene Details
Entrez Gene ID: | 10718 |
Gene Name: | neuregulin 3 |
Gene Aliases: |
HRG3, pro-NRG3 |
Location: |
Chr.10:81875298-82989979 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551669 | Chr.10:81374678 - 82080786 on Build GRCh38 | Gain |
|
| esv3891873 | Chr.10:81025608 - 82340753 on Build GRCh38 | Gain |
|
| nsv831925 | Chr.10:81771475 - 81938851 on Build GRCh38 | Gain |
|
| nsv551670 | Chr.10:81545860 - 81972366 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map