Assay Details
Target Gene Details
Entrez Gene ID: | 9200 |
Gene Name: | 3-hydroxyacyl-CoA dehydratase 1 |
Gene Aliases: |
CAP, PTPLA |
Location: |
Chr.10:17589959-17617374 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACD1 | NM_014241.3 | NP_055056.3 | ||
| XM_005252641.4 | XP_005252698.1 | |||
| AF114494.1 | AAF21976.1 | |||
| BC010353.1 | AAH10353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2669454 | Chr.10:17590796 - 17593440 on Build GRCh38 | Deletion |
|
| esv3622493 | Chr.10:17590956 - 17593604 on Build GRCh38 | Loss |
|
| esv3578665 | Chr.10:17591060 - 17596181 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map