Assay Details
Target Gene Details
Entrez Gene ID: | 102724341 |
Gene Name: | NEURL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:103479603-103517442 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEURL1-AS1 | NR_120675.1 | |||
| AA399019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524966 | Chr.10:103441820 - 103717405 on Build GRCh38 | Loss |
|
| nsv518987 | Chr.10:103434329 - 103484808 on Build GRCh38 | Loss |
|
| dgv966n100 | Chr.10:103385173 - 103553473 on Build GRCh38 | Gain |
|
| nsv525801 | Chr.10:103480929 - 103484808 on Build GRCh38 | Loss |
|
| esv3577694 | Chr.10:103391276 - 103553473 on Build GRCh38 | Gain |
|

Genomic Map