Assay Details
Target Gene Details
Entrez Gene ID: | 83938 |
Gene Name: | chromosome 10 open reading frame 11 |
Gene Aliases: |
CDA017 |
Location: |
Chr.10:75431646-76559220 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf11 | NM_001305581.1 | NP_001292510.1 | ||
| NR_131178.1 | ||||
| XM_011540256.2 | XP_011538558.1 | |||
| XM_011540257.2 | XP_011538559.1 | |||
| XM_017016759.1 | XP_016872248.1 | |||
| XM_017016760.1 | XP_016872249.1 | |||
| XM_017016761.1 | XP_016872250.1 | |||
| BM720401.1 | ||||
| BX105111.1 | ||||
| CR933639.1 | ||||
| HY006640.1 |
Target Gene Details
Entrez Gene ID: | 105378367 |
Gene Name: | uncharacterized LOC105378367 |
Gene Aliases: |
- |
Location: |
Chr.10:75712455-75743848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105378367 | NR_132791.1 | |||
| HY007587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831914 | Chr.10:75590441 - 75760988 on Build GRCh38 | Gain |
|
| esv2759768 | Chr.10:75475179 - 76010691 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74433057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map