Assay Details
Target Gene Details
Entrez Gene ID: | 51196 |
Gene Name: | phospholipase C epsilon 1 |
Gene Aliases: |
NPHS3, PLCE, PPLC |
Location: |
Chr.10:93993989-94329447 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCE1 | NM_001165979.2 | NP_001159451.1 | ||
| NM_001288989.1 | NP_001275918.1 | |||
| NM_016341.3 | NP_057425.3 | |||
| XM_006717885.3 | XP_006717948.1 | |||
| XM_006717888.3 | XP_006717951.1 | |||
| XM_006717889.3 | XP_006717952.1 | |||
| XM_006717890.2 | XP_006717953.1 | |||
| XM_011539849.2 | XP_011538151.1 | |||
| XM_011539850.2 | XP_011538152.1 | |||
| XM_011539851.2 | XP_011538153.1 | |||
| XM_011539852.2 | XP_011538154.1 | |||
| XM_017016310.1 | XP_016871799.1 | |||
| XM_017016311.1 | XP_016871800.1 | |||
| XM_017016312.1 | XP_016871801.1 | |||
| AB040949.2 | ||||
| AF170071.1 | AAG28341.1 | |||
| AF190642.2 | AAG17145.2 | |||
| AK289852.1 | ||||
| AK297779.1 | ||||
| BC140705.1 | ||||
| BC144286.1 | ||||
| BC151854.1 | ||||
| BX491865.1 |
Target Gene Details
Entrez Gene ID: | 101927049 |
Gene Name: | PLCE1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.10:94081950-94108794 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCE1-AS2 | NR_120615.1 | |||
| NR_120616.1 | ||||
| AK098548.1 | ||||
| HY019250.1 |
More Information
Set Membership: |
|

Genomic Map