Assay Details
Target Gene Details
Entrez Gene ID: | 2849 |
Gene Name: | G protein-coupled receptor 26 |
Gene Aliases: |
- |
Location: |
Chr.10:123666355-123697399 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR26 | NM_153442.3 | NP_703143.1 | ||
| AJ505757.1 | CAD44281.1 | |||
| BC112011.1 | AAI12012.1 | |||
| BC113462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552276 | Chr.10:123668548 - 123688189 on Build GRCh38 | Loss |
|
| nsv832011 | Chr.10:123590498 - 123760281 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map