Assay Details
Target Gene Details
Entrez Gene ID: | 5599 |
Gene Name: | mitogen-activated protein kinase 8 |
Gene Aliases: |
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c |
Location: |
Chr.10:48306639-48439360 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK8 | NM_001278547.1 | NP_001265476.1 | ||
| NM_001323302.1 | NP_001310231.1 | |||
| NM_001323320.1 | NP_001310249.1 | |||
| NM_001323321.1 | NP_001310250.1 | |||
| NM_001323322.1 | NP_001310251.1 | |||
| NM_001323323.1 | NP_001310252.1 | |||
| NM_001323324.1 | NP_001310253.1 | |||
| NM_001323325.1 | NP_001310254.1 | |||
| NM_001323326.1 | NP_001310255.1 | |||
| NM_001323327.1 | NP_001310256.1 | |||
| NM_001323328.1 | NP_001310257.1 | |||
| NM_001323329.1 | NP_001310258.1 | |||
| NM_001323330.1 | NP_001310259.1 | |||
| NM_001323331.1 | NP_001310260.1 | |||
| NM_139046.3 | NP_620634.1 | |||
| NM_139049.3 | NP_620637.1 | |||
| NR_136583.1 | ||||
| NR_136584.1 | ||||
| NR_136585.1 | ||||
| AK292523.1 | ||||
| DA159335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6587 | Chr.10:48318286 - 48363761 on Build GRCh38 | Deletion |
|
| nsv467190 | Chr.10:48194041 - 48368835 on Build GRCh38 | Gain |
|
| esv3623153 | Chr.10:48355632 - 48366278 on Build GRCh38 | Loss |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map