Assay Details
Target Gene Details
Entrez Gene ID: | 2894 |
Gene Name: | glutamate ionotropic receptor delta type subunit 1 |
Gene Aliases: |
GluD1 |
Location: |
Chr.10:85599555-86366493 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1 | NM_017551.2 | NP_060021.1 | ||
| XM_011539720.2 | XP_011538022.1 | |||
| AK127168.1 | ||||
| AK293104.1 | ||||
| BC039263.1 | AAH39263.1 | |||
| DR001156.1 |
Target Gene Details
Entrez Gene ID: | 442911 |
Gene Name: | microRNA 346 |
Gene Aliases: |
MIRN346, hsa-mir-346, miR-346, miRNA346 |
Location: |
Chr.10:86264694-86264788 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR346 | NR_029907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831938 | Chr.10:86216648 - 86358426 on Build GRCh38 | Gain |
|
| nsv551773 | Chr.10:86222102 - 86302046 on Build GRCh38 | Loss |
|
| nsv1051761 | Chr.10:86238768 - 86305399 on Build GRCh38 | Loss |
|
| nsv831936 | Chr.10:86107087 - 86297632 on Build GRCh38 | Loss |
|
| nsv520914 | Chr.10:86236694 - 86302046 on Build GRCh38 | Loss |
|
| esv3579031 | Chr.10:86237805 - 86303891 on Build GRCh38 | Loss |
|
| nsv1052427 | Chr.10:85234813 - 86270793 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map