Assay Details
Target Gene Details
Entrez Gene ID: | 5092 |
Gene Name: | pterin-4 alpha-carbinolamine dehydratase 1 |
Gene Aliases: |
DCOH, PCBD, PCD, PHS |
Location: |
Chr.10:70882280-70888786 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCBD1 | NM_001323004.1 | NP_001309933.1 | ||
| AA397961.1 | ||||
| BM724638.1 | ||||
| HY292808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551495 | Chr.10:70852412 - 70891937 on Build GRCh38 | Loss |
|
| nsv1043245 | Chr.10:70290210 - 70915897 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map