Assay Details
Target Gene Details
Entrez Gene ID: | 2894 |
Gene Name: | glutamate ionotropic receptor delta type subunit 1 |
Gene Aliases: |
GluD1 |
Location: |
Chr.10:85599555-86366493 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRID1 | NM_017551.2 | NP_060021.1 | ||
| XM_011539720.2 | XP_011538022.1 | |||
| AB033046.1 | BAA86534.1 | |||
| AK127168.1 | ||||
| AK293104.1 | ||||
| AK302192.1 | ||||
| BC039263.1 | AAH39263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891878 | Chr.10:85560815 - 85628886 on Build GRCh38 | Loss |
|
| nsv1052427 | Chr.10:85234813 - 86270793 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map