Assay Details
Target Gene Details
Entrez Gene ID: | 159371 |
Gene Name: | solute carrier family 35 member G1 |
Gene Aliases: |
C10orf60, TMEM20 |
Location: |
Chr.10:93893951-93914265 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC35G1 | XM_011539373.2 | XP_011537675.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
