Assay Details
Target Gene Details
Entrez Gene ID: | 22986 |
Gene Name: | sortilin related VPS10 domain containing receptor 3 |
Gene Aliases: |
SORCS |
Location: |
Chr.10:104641101-105265242 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS3 | NM_014978.2 | NP_055793.1 | ||
| AB028982.1 | BAA83011.2 | |||
| BC117417.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579100 | Chr.10:104573121 - 104983037 on Build GRCh38 | Loss |
|
| nsv831972 | Chr.10:104523290 - 104689830 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74155013] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map