Assay Details
Target Gene Details
Entrez Gene ID: | 57118 |
Gene Name: | calcium/calmodulin dependent protein kinase ID |
Gene Aliases: |
CKLiK, CaM-K1, CaMKID |
Location: |
Chr.10:12349514-12835545 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK1D | NM_020397.3 | NP_065130.1 | ||
| NM_153498.3 | NP_705718.1 | |||
| XM_006717482.3 | 1 | 52 | XP_006717545.1 | |
| XM_006717483.3 | XP_006717546.1 | |||
| XM_017016438.1 | 1 | 50 | XP_016871927.1 | |
| AB081726.1 | BAC19846.1 | |||
| AK289393.1 | ||||
| BU542765.1 | 1 | 84 | ||
| HY168807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112574 | Chr.10:12348901 - 12349701 on Build GRCh38 | Deletion |
|
| nsv1069022 | Chr.10:12348900 - 12349701 on Build GRCh38 | Deletion |
|
| esv3622363 | Chr.10:12200775 - 12828318 on Build GRCh38 | Gain |
|
| esv2676569 | Chr.10:12231868 - 12393054 on Build GRCh38 | Deletion |
|
| nsv951250 | Chr.10:12348602 - 12349701 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map