Assay Details
Target Gene Details
Entrez Gene ID: | 4153 |
Gene Name: | mannose binding lectin 2 |
Gene Aliases: |
COLEC1, HSMBPC, MBL, MBL2D, MBP, MBP-C, MBP1, MBPD |
Location: |
Chr.10:52764977-52772845 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBL2 | NM_000242.2 | NP_000233.1 | ||
| XM_006717861.3 | XP_006717924.1 | |||
| XM_011539816.2 | XP_011538118.1 | |||
| AF360991.1 | AAK52907.1 | |||
| AY826184.1 | AAV80468.1 | |||
| BC069338.1 | AAH69338.1 | |||
| BC096179.3 | AAH96179.1 | |||
| BC096180.3 | ||||
| BC096181.3 | ||||
| BC096182.3 | ||||
| X15422.1 | CAA33462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041917 | Chr.10:52741572 - 55682853 on Build GRCh38 | Loss |
|
| dgv1140n54 | Chr.10:52764898 - 52772139 on Build GRCh38 | Loss |
|
| nsv437673 | Chr.10:52765749 - 52776791 on Build GRCh38 | Loss |
|
| dgv855n100 | Chr.10:52759208 - 52777917 on Build GRCh38 | Gain |
|
| dgv854n100 | Chr.10:52757467 - 52785830 on Build GRCh38 | Gain |
|
| esv3578899 | Chr.10:52761345 - 52773600 on Build GRCh38 | Loss |
|
| nsv831876 | Chr.10:52732751 - 52889632 on Build GRCh38 | Gain |
|
| nsv1051777 | Chr.10:52761330 - 52802372 on Build GRCh38 | Gain |
|
| dgv856n100 | Chr.10:52769592 - 52789557 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map