Assay Details
Target Gene Details
Entrez Gene ID: | 94134 |
Gene Name: | Rho GTPase activating protein 12 |
Gene Aliases: |
- |
Location: |
Chr.10:31805398-31928876 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP12 | NM_001270695.1 | 19 | 4456 | NP_001257624.1 |
| NM_001270696.1 | 18 | 4381 | NP_001257625.1 | |
| NM_001270697.1 | 19 | 4330 | NP_001257626.1 | |
| NM_001270698.1 | 17 | 4240 | NP_001257627.1 | |
| NM_001270699.1 | 16 | 4082 | NP_001257628.1 | |
| NM_018287.6 | 20 | 4471 | NP_060757.4 | |
| XM_005252644.1 | 18 | 4315 | XP_005252701.1 | |
| XM_011519761.1 | 18 | 4249 | XP_011518063.1 | |
| XM_017016954.1 | 16 | 4159 | XP_016872443.1 | |
| AL137485.1 | 11 | 2909 | CAB70766.1 | |
| AL834250.2 | 16 | 4006 | CAD38926.2 | |
| BM693575.1 | 1 | 74 | ||
| BX641115.1 | 2 | 3825 | CAE46053.1 |
More Information
Set Membership: |
|

Genomic Map