Assay Details
Target Gene Details
Entrez Gene ID: | 2572 |
Gene Name: | glutamate decarboxylase 2 |
Gene Aliases: |
GAD65 |
Location: |
Chr.10:26216307-26304562 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAD2 | NM_000818.2 | NP_000809.1 | ||
| NM_001134366.1 | NP_001127838.1 | |||
| AJ251501.1 | CAB62572.1 | |||
| BC126327.1 | ||||
| BC126329.1 | ||||
| M74826.1 | AAA58491.1 | |||
| M81882.1 | AAA62367.1 | |||
| X69936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576316 | Chr.10:26224643 - 26386823 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map