Assay Details
Target Gene Details
Entrez Gene ID: | 3778 |
Gene Name: | potassium calcium-activated channel subfamily M alpha 1 |
Gene Aliases: |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
Location: |
Chr.10:76869601-77637819 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Gene Details
Entrez Gene ID: | 101929310 |
Gene Name: | KCNMA1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.10:77140831-77150100 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMA1-AS2 | NR_120654.1 | 3 | 290 | |
| AA983877.1 | 1 | 158 |
More Information
Set Membership: |
|

Genomic Map