Assay Details
Target Gene Details
Entrez Gene ID: | 51196 |
Gene Name: | phospholipase C epsilon 1 |
Gene Aliases: |
NPHS3, PLCE, PPLC |
Location: |
Chr.10:93993989-94329447 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCE1 | NM_001288989.1 | NP_001275918.1 | ||
| NM_016341.3 | NP_057425.3 | |||
| XM_006717885.3 | XP_006717948.1 | |||
| XM_006717888.3 | XP_006717951.1 | |||
| XM_006717889.3 | XP_006717952.1 | |||
| XM_011539849.2 | XP_011538151.1 | |||
| XM_011539851.2 | XP_011538153.1 | |||
| XM_011539852.2 | XP_011538154.1 | |||
| XM_017016310.1 | XP_016871799.1 | |||
| XM_017016311.1 | XP_016871800.1 | |||
| AB040949.2 | ||||
| BC151854.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72812649] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map