Assay Details
Target Gene Details
Entrez Gene ID: | 51008 |
Gene Name: | activating signal cointegrator 1 complex subunit 1 |
Gene Aliases: |
ASC1p50, CGI-18, SMABF2, p50 |
Location: |
Chr.10:72096032-72217134 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891857 | Chr.10:72086126 - 72148450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map