Assay Details
Target Gene Details
Entrez Gene ID: | 29119 |
Gene Name: | catenin alpha 3 |
Gene Aliases: |
ARVD13, VR22 |
Location: |
Chr.10:65912518-67696217 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNA3 | NM_001127384.2 | 18 | 4871 | NP_001120856.1 |
| NM_013266.3 | 18 | 4918 | NP_037398.2 | |
| XM_017016151.1 | 19 | 4913 | XP_016871640.1 | |
| XM_017016152.1 | 21 | 5210 | XP_016871641.1 | |
| XM_017016153.1 | 19 | 4921 | XP_016871642.1 | |
| XM_017016154.1 | 14 | 4289 | XP_016871643.1 | |
| XM_017016155.1 | 14 | 4299 | XP_016871644.1 | |
| XM_017016156.1 | 14 | 4176 | XP_016871645.1 | |
| XM_017016157.1 | 12 | 4326 | XP_016871646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047351 | Chr.10:65563025 - 66683052 on Build GRCh38 | Gain |
|
| nsv1037793 | Chr.10:65517853 - 66063168 on Build GRCh38 | Loss |
|
| esv1004637 | Chr.10:65908206 - 65919037 on Build GRCh38 | Deletion |
|
| nsv551286 | Chr.10:65550888 - 66450664 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map