Assay Details
Target Gene Details
Entrez Gene ID: | 8644 |
Gene Name: | aldo-keto reductase family 1, member C3 |
Gene Aliases: |
DD3, DDX, HA1753, HAKRB, HAKRe, HSD17B5, PGFS, hluPGFS |
Location: |
Chr.10:5048766-5107686 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1C3 | NM_001253908.1 | NP_001240837.1 | ||
| AK296829.1 | ||||
| DQ269985.1 | ABB76132.1 | |||
| H79159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891742 | Chr.10:4974605 - 5101845 on Build GRCh38 | Loss |
|
| nsv1161924 | Chr.10:5067963 - 5090996 on Build GRCh38 | Duplication |
|
| esv2763913 | Chr.10:4918829 - 5279444 on Build GRCh38 | Gain+Loss |
|
| nsv549881 | Chr.10:4859606 - 5277716 on Build GRCh38 | Loss |
|
| nsv1036566 | Chr.10:4971312 - 5219463 on Build GRCh38 | Loss |
|
| nsv549882 | Chr.10:4901326 - 5161901 on Build GRCh38 | Loss |
|
| nsv1045155 | Chr.10:4830047 - 5169355 on Build GRCh38 | Loss |
|
| esv3622214 | Chr.10:4972626 - 5103318 on Build GRCh38 | Loss |
|
| nsv549883 | Chr.10:4983904 - 5233799 on Build GRCh38 | Loss |
|
| nsv1050246 | Chr.10:4885236 - 5169355 on Build GRCh38 | Gain |
|
| nsv1039386 | Chr.10:4966191 - 5344882 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111537719] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map