Assay Details
Target Gene Details
Entrez Gene ID: | 84795 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 2 |
Gene Aliases: |
C10orf33, FP3420 |
Location: |
Chr.10:98383565-98415221 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD2 | NM_032709.2 | NP_116098.2 | ||
| XM_011540293.2 | XP_011538595.1 | |||
| XM_011540301.2 | XP_011538603.1 | |||
| XM_017016835.1 | XP_016872324.1 | |||
| XM_017016838.1 | XP_016872327.1 | |||
| XM_017016839.1 | XP_016872328.1 | |||
| XM_017016840.1 | XP_016872329.1 | |||
| XM_017016841.1 | XP_016872330.1 | |||
| AK022259.1 | 1 | 1660 | ||
| AK074429.1 | ||||
| AK075265.1 | BAC11507.1 | |||
| AK123713.1 | ||||
| AK225637.1 | ||||
| BC006131.1 | AAH06131.1 |
More Information
Set Membership: |
|

Genomic Map