Assay Details
Target Gene Details
Entrez Gene ID: | 10718 |
Gene Name: | neuregulin 3 |
Gene Aliases: |
HRG3, pro-NRG3 |
Location: |
Chr.10:81875298-82989979 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Gene Details
Entrez Gene ID: | 101929590 |
Gene Name: | NRG3 antisense RNA 1 |
Gene Aliases: |
C10orf100, Em:AC010157.1 |
Location: |
Chr.10:82229014-82232920 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRG3-AS1 | CF272718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551704 | Chr.10:82136504 - 82268652 on Build GRCh38 | Loss |
|
| esv3891873 | Chr.10:81025608 - 82340753 on Build GRCh38 | Gain |
|
| esv3891875 | Chr.10:82171786 - 82294496 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74615677] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map