Assay Details
Target Gene Details
Entrez Gene ID: | 100506733 |
Gene Name: | FAM170B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:49121839-49151547 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM170B-AS1 | NR_038973.1 | |||
| AK093055.1 |
Target Gene Details
Entrez Gene ID: | 196740 |
Gene Name: | V-set and transmembrane domain containing 4 |
Gene Aliases: |
C10orf72 |
Location: |
Chr.10:49014245-49129118 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM4 | XM_017015825.1 | XP_016871314.1 | ||
| XM_017015826.1 | XP_016871315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623164 | Chr.10:49113706 - 49131289 on Build GRCh38 | Loss |
|
| esv2761593 | Chr.10:49068767 - 49128084 on Build GRCh38 | Gain |
|
| nsv831866 | Chr.10:48957135 - 49127231 on Build GRCh38 | Gain+Loss |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map