Assay Details
Target Gene Details
Entrez Gene ID: | 81894 |
Gene Name: | solute carrier family 25 member 28 |
Gene Aliases: |
MFRN2, MRS3/4, MRS4L, NPD016 |
Location: |
Chr.10:99610518-99659329 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A28 | NM_031212.3 | NP_112489.3 | ||
| XM_005270208.1 | XP_005270265.1 | |||
| XM_005270211.4 | XP_005270268.1 | |||
| XM_006718005.3 | XP_006718068.1 | |||
| XM_006718006.3 | XP_006718069.1 | |||
| XM_011540239.2 | XP_011538541.1 | |||
| XM_011540240.1 | 1 | 924 | XP_011538542.1 | |
| XM_011540244.2 | XP_011538546.1 | |||
| XM_017016741.1 | 1 | 924 | XP_016872230.1 | |
| XM_017016742.1 | XP_016872231.1 | |||
| AF327402.1 | AAK49519.1 | |||
| AF327403.1 | AAK49520.1 | |||
| AJ303077.1 | CAC27996.1 | |||
| AJ303078.1 | CAC27997.1 | |||
| AK094405.1 | 1 | 925 | ||
| BC047312.1 | AAH47312.1 | |||
| BC058937.1 | AAH58937.1 | |||
| BC076399.1 | AAH76399.1 | |||
| BC094821.1 | AAH94821.1 | |||
| CD672723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7515 | Chr.10:99606658 - 99639439 on Build GRCh38 | Insertion |
|
| nsv831958 | Chr.10:99451304 - 99651424 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map