Assay Details
Target Gene Details
Entrez Gene ID: | 3257 |
Gene Name: | HPS1, biogenesis of lysosomal organelles complex 3 subunit 1 |
Gene Aliases: |
BLOC3S1, HPS |
Location: |
Chr.10:98415068-98446963 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPS1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524427 | Chr.10:98408615 - 98452617 on Build GRCh38 | Loss |
|
| nsv515723 | Chr.10:98388419 - 98435072 on Build GRCh38 | Loss |
|
| nsv831955 | Chr.10:98402137 - 98592058 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79854246] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map