Assay Details
Target Gene Details
Entrez Gene ID: | 340895 |
Gene Name: | MAM and LDL receptor class A domain containing 1 |
Gene Aliases: |
C10orf112, DIET1, bA265G8.2 |
Location: |
Chr.10:19046931-19734478 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MALRD1 | XM_011519453.2 | XP_011517755.1 | ||
| XM_011519454.1 | XP_011517756.1 | |||
| XM_011519455.2 | XP_011517757.1 | |||
| XM_017016183.1 | XP_016871672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv682n100 | Chr.10:19030782 - 19058141 on Build GRCh38 | Loss |
|
| nsv528472 | Chr.10:19007821 - 19096319 on Build GRCh38 | Gain |
|
| nsv516123 | Chr.10:19031864 - 19056050 on Build GRCh38 | Loss |
|
| nsv550126 | Chr.10:18981139 - 19283800 on Build GRCh38 | Gain |
|
| esv2665204 | Chr.10:18927737 - 19089058 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79980591] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map