Assay Details
Target Gene Details
Entrez Gene ID: | 83592 |
Gene Name: | aldo-keto reductase family 1 member E2 |
Gene Aliases: |
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR |
Location: |
Chr.10:4824554-4908421 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1E2 | XM_011519715.2 | 1 | 316 | XP_011518017.1 |
| XM_011519718.2 | 1 | 316 | XP_011518020.1 | |
| XM_011519719.2 | 1 | 316 | XP_011518021.1 | |
| XM_011519720.2 | 1 | 315 | XP_011518022.1 | |
| XM_011519722.2 | 1 | 316 | XP_011518024.1 | |
| XM_011519724.2 | 1 | 315 | XP_011518026.1 | |
| XM_011519725.2 | 1 | 313 | XP_011518027.1 | |
| XM_011519729.2 | 1 | 313 | XP_011518031.1 | |
| XM_017016743.1 | 1 | 317 | XP_016872232.1 | |
| XM_017016744.1 | 1 | 316 | XP_016872233.1 |
More Information
Set Membership: |
|

Genomic Map