Assay Details
Target Gene Details
Entrez Gene ID: | 8036 |
Gene Name: | SHOC2, leucine rich repeat scaffold protein |
Gene Aliases: |
SIAA0862, SOC2, SUR8 |
Location: |
Chr.10:110919370-111013667 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHOC2 | NM_001269039.2 | NP_001255968.1 | ||
| NM_001324336.1 | NP_001311265.1 | |||
| NM_001324337.1 | NP_001311266.1 | |||
| NM_007373.3 | NP_031399.2 | |||
| NR_136749.1 | ||||
| XM_017016704.1 | XP_016872193.1 | |||
| AB020669.1 | ||||
| AF054828.1 | AAC25698.1 | |||
| AK292833.1 | ||||
| AK293271.1 | ||||
| BX649055.1 | ||||
| DB451165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035602 | Chr.10:110876295 - 111021655 on Build GRCh38 | Loss |
|
| nsv831985 | Chr.10:110892331 - 111043345 on Build GRCh38 | Gain+Loss |
|
| nsv518667 | Chr.10:110933044 - 110937483 on Build GRCh38 | Loss |
|
| dgv974n100 | Chr.10:110924064 - 111035401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map