Assay Details
Target Gene Details
Entrez Gene ID: | 84795 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 2 |
Gene Aliases: |
C10orf33, FP3420 |
Location: |
Chr.10:98383565-98415221 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYROXD2 | NM_032709.2 | NP_116098.2 | ||
| XM_011540293.2 | XP_011538595.1 | |||
| XM_017016835.1 | XP_016872324.1 | |||
| XM_017016836.1 | XP_016872325.1 | |||
| XM_017016837.1 | XP_016872326.1 | |||
| XM_017016838.1 | 4 | 1990 | XP_016872327.1 | |
| XM_017016839.1 | XP_016872328.1 | |||
| AK074429.1 | ||||
| AK075265.1 | BAC11507.1 | |||
| AK225637.1 | ||||
| AK295699.1 | ||||
| BC006131.1 | AAH06131.1 | |||
| DC361966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv515723 | Chr.10:98388419 - 98435072 on Build GRCh38 | Loss |
|
| nsv831955 | Chr.10:98402137 - 98592058 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map