Assay Details
Target Gene Details
Entrez Gene ID: | 256297 |
Gene Name: | pancreas specific transcription factor, 1a |
Gene Aliases: |
PACA, PAGEN2, PTF1-p48, bHLHa29 |
Location: |
Chr.10:23192531-23194252 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTF1A | NM_178161.2 | 1 | 633 | NP_835455.1 |
| BM055347.1 | 1 | 206 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825302 | Chr.10:23168093 - 23208400 on Build GRCh38 | Gain |
|
| nsv825301 | Chr.10:23151675 - 23216917 on Build GRCh38 | Gain |
|
| nsv1068293 | Chr.10:23189170 - 23194971 on Build GRCh38 | Deletion |
|
| nsv831810 | Chr.10:23156718 - 23316341 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map