Assay Details
Target Gene Details
Entrez Gene ID: | 8029 |
Gene Name: | cubilin |
Gene Aliases: |
IFCR, MGA1, gp280 |
Location: |
Chr.10:16823966-17130492 on Build GRCh38 |
Assay Gene Location: | Within Intron 63 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUBN | NM_001081.3 | NP_001072.2 | ||
| XM_011519709.2 | XP_011518011.1 | |||
| XM_011519710.2 | XP_011518012.1 | |||
| XM_011519711.2 | XP_011518013.1 | |||
| AB209880.1 | BAD93117.1 | |||
| AF034611.1 | AAC82612.1 | |||
| AK075228.1 | ||||
| AK310404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470924 | Chr.10:16864509 - 16951578 on Build GRCh38 | Gain |
|
| esv3891765 | Chr.10:16809409 - 16977014 on Build GRCh38 | Gain |
|
| nsv550020 | Chr.10:16672467 - 16954685 on Build GRCh38 | Gain |
|
| nsv831800 | Chr.10:16765669 - 16949419 on Build GRCh38 | Gain |
|
| nsv1046088 | Chr.10:16786830 - 16906310 on Build GRCh38 | Gain |
|
| esv3622477 | Chr.10:16808207 - 16934688 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map