Assay Details
Target Gene Details
Entrez Gene ID: | 5309 |
Gene Name: | paired like homeodomain 3 |
Gene Aliases: |
ASMD, ASOD, CTPP4, CTRCT11, PTX3 |
Location: |
Chr.10:102230189-102241474 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITX3 | NM_005029.3 | NP_005020.1 | ||
| BC011642.2 | AAH11642.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs148311798] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map